Labeling tapes, TX
Label printer tape, Polyester, Thermal transfer, sort/hvid, PT-7000/8000, 6 mm×15 m
Katalog # BROTTX-211
Leverandør: BROTHER
Gå til:
Product Details & Documents
Specifikationer
- Printer:PT-7000/8000
- Printing method:Thermal transfer
- Farve:sort/hvid
- Beskrivelse:Label printer tape
- Dimensioner:6 mm×15 m
- Materiale:Polyester
- Seq:01