Labeling tapes, TX
Label printer tape, Polyester, Thermal transfer, sort/hvid, PT-7000/8000, 18 mm×15 m
Catalog # BROTTX-345
Supplier: BROTHER
Jump to:
Product Details & Documents
Specifications
- Printer:PT-7000/8000
- Printing method:Thermal transfer
- Farve:sort/hvid
- Beskrivelse:Label printer tape
- Dimensioner:18 mm×15 m
- Materiale:Polyester
- Seq:14