Labeling tapes, TX
Label printer tape, Polyester, Thermal transfer, hvid/blå, PT-7000/8000, 24 mm×15 m
Katalog # BROTTX-253
Leverandør: BROTHER
Gå til:
Product Details & Documents
Specifikationer
- Printer:PT-7000/8000
- Printing method:Thermal transfer
- Farve:hvid/blå
- Beskrivelse:Label printer tape
- Dimensioner:24 mm×15 m
- Materiale:Polyester
- Seq:20