Reading pocket magnifiers
Håndholdt sammenfoldelig lup i brunt læderetui, 3,5×, 60 mm
Catalog # ESCH1740460
Supplier: ESCHENBACH OPTIK
Jump to:
Product Details & Documents
These handheld folding magnifiers with classic design fit easily into any handbag or pocket.
- Many different colours and sizes
- Anti-scratch coating for both models
- Black or brown leather case available
Specifications
- Ø:60 mm
- Forstørrelse:3,5×
- Beskrivelse:Håndholdt sammenfoldelig lup i brunt læderetui