Indslagslupper
Folding double magnifier, biconvex, 4×; 6×, 30 mm
Katalog # 630-2599
Leverandør: ESCHENBACH OPTIK
Gå til:
Product Details & Documents
Swivelling black metallic magnifier with loop, lens frame made from black painted metal.
- Protective case can serve as a handle
- Provide distortion-free images
- Chrome plated metal
Specifikationer
- Ø:30 mm
- Forstørrelse:4×; 6×
- Beskrivelse:Folding double magnifier, biconvex
- Seq:005