Orders LinkContactUsLinkComponent
Glass beads
Glass beads
Glass beads, solid, 6 mm
Katalog # 201-0280
Glass beads
Katalog # 201-0280
Glass beads
Glass beads
Glass beads, solid, 6 mm
Price
262,00 kr
Log ind for tilgængelighed
1 * 1 kg
Product Details & Documents
Solid glass.
For packing distillation columns.
Specifikationer
  • Ø:
    6 mm
  • Beskrivelse:
    Glass beads, solid
  • Seq:
    07