Orders LinkContactUsLinkComponent
Glass beads
Glass beads
Glass beads, solid, 4 mm
Catalog # 201-0278
Glass beads
Catalog # 201-0278
Glass beads
Glass beads
Glass beads, solid, 4 mm
Price
263,00 kr
Log in for availability
1 * 1 kg
Product Details & Documents
Solid glass.
For packing distillation columns.
Specifications
  • Ø:
    4 mm
  • Beskrivelse:
    Glass beads, solid
  • Seq:
    04