Cover glasses
Cover glasses, square, without cords and blisters, 24×60 mm, #1
Katalog # KNITVD12460Y100A
Leverandør: KNITTEL
Gå til:
Product Details & Documents
Borosilicate glass of hydrolytic class 1 cover glasses with precise cutting.
: ISO 8255/1
: 100 pieces per box - 1000 pieces per carton.
Specifikationer
- Antal:#1
- Beskrivelse:Cover glasses, square, without cords and blisters
- L×B:24×60 mm