Single channel pipettes, variable volume, MLA Digital Adjustable
200 - 1000 µl, ±1 %, <0,50 - <0,60 %, grå
Katalog # VIST1138
Leverandør: VISTALAB TECHNOLOGIES
Single channel pipettes, variable volume, MLA Digital Adjustable
Katalog # VIST1138
Leverandør: VISTALAB TECHNOLOGIES
Gå til:
Product Details & Documents
Digital adjustable pipettes are an ideal choice for laboratories that perform testing requiring multiple or non standard volumes. They are made of engineered aluminum alloy and stainless steel for maximum durability and performance.
- Three models to cover volume range of 10 to 1000 μl
- Lower spring forces to minimise thumb muscle activity
- Simple dial-in settings with 'click stops' provide fast and easy volume selection
- Streamlined shape and smooth, singlestroke operation make these pipettes easy to use
All pipettes are engraved with serial numbers to facilitate improved traceability and calibration recordkeeping.
Specifikationer
- Farvekode:grå
- Volumen:200 - 1000 µl
- Seq:03
- Nøjagtighed:±1 %
- Usikkerhed:<0,50 - <0,60 %
Accessories
Produktinformation
- Katalog #TilgængelighedPrice
- MLA pipette nozzle insert, white
Produktinformation
- Katalog #TilgængelighedPrice
- Seal kit for 10 - 50 µl Digital pipette
Produktinformation
- Katalog #TilgængelighedPrice
- Seal kit for 50 - 200 µl Digital pipetteSeal kit for 50 - 200 µl Digital pipetteVIST9092Log ind for tilgængelighed1 * 1 Items - 225,00 kr
Produktinformation
- Katalog #TilgængelighedPrice
- Seal kit for 200 - 1000 µl Digital pipetteSeal kit for 200 - 1000 µl Digital pipetteVIST9094Log ind for tilgængelighed1 * 1 Items - 225,00 kr
Produktinformation
- Katalog #TilgængelighedPrice
- MLA pipette stand for six pipettes
Produktinformation
- Katalog #TilgængelighedPrice
- Seal lubricant (replacement for seal kit)Seal lubricant (replacement for seal kit)VIST9030Log ind for tilgængelighed1 * 1 Items - 141,00 kr
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



