Pipette tips
In box, 5 ml, -, 100 tips/box
Catalog # VIST9048
Supplier: VISTALAB TECHNOLOGIES
Jump to:
Product Details & Documents
These tips to maximise the exceptional performance and benefits of ovation and MLA pipettes. All VistaLab pipettes are calibrated with VistaLab tips to provide a perfect fit and deliver reliable performance every time.
Specifications
- Pakket:100 tips/box
- Steril:-
- Volumen:5 ml
- Seq:23
- Spidstype:In box