Orders LinkContactUsLinkComponent
Bottle top tilt measures
Bottle top tilt measures
50 ml
Katalog # 613-8720
Leverandør:   witeg Labortechnik
Bottle top tilt measures
Katalog # 613-8720
Leverandør:  witeg Labortechnik
Bottle top tilt measures
Bottle top tilt measures
50 ml
Price
725,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Glass, with normal-shaped core.
  • For repetitive dispensing of the same volume
  • Can bleed with all bottles with NS 29/32
Specifikationer
  • Kapacitet:
    50 ml