Orders LinkContactUsLinkComponent
Bottle top tilt measures
Bottle top tilt measures
100 ml
  613-8721
Bottle top tilt measures
  613-8721
Bottle top tilt measures
Bottle top tilt measures
100 ml
845,00 kr
1 * 1 Items
Glass, with normal-shaped core.
  • For repetitive dispensing of the same volume
  • Can bleed with all bottles with NS 29/32
  • Kapacitet:
    100 ml