Bottle top tilt measures
10 ml
Katalog # 613-8714
Leverandør: witeg Labortechnik
Gå til:
Product Details & Documents
Glass, with normal-shaped core.
- For repetitive dispensing of the same volume
- Can bleed with all bottles with NS 29/32
Specifikationer
- Kapacitet:10 ml