Orders LinkContactUsLinkComponent
Bottle top tilt measures
Bottle top tilt measures
2 ml
Katalog # 613-8712
Leverandør:   witeg Labortechnik
Bottle top tilt measures
Katalog # 613-8712
Leverandør:  witeg Labortechnik
Bottle top tilt measures
Bottle top tilt measures
2 ml
Price
434,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Glass, with normal-shaped core.
  • For repetitive dispensing of the same volume
  • Can bleed with all bottles with NS 29/32
Specifikationer
  • Kapacitet:
    2 ml