Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Dropping bottles, glass

Dropping bottles, glass

100 ml, 110 mm, 47 mm
Catalog # 215-3753
Dropping bottles, glass
Catalog # 215-3753
Supplier:  
Dropping bottles, glass
Dropping bottles, glass

100 ml, 110 mm, 47 mm

Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
[EN]PIPETTE KIT BLUE FOR 100ML BOTTLE 1 * 100 Items Catalog #215-3828

Product 215-3828 is similar to 215-3753 but manufacturer and specifications may vary.

Product Details & Documents

Glass, amber, with screw cap, pipette and rubber suction bulb.

  • Good chemical and physical properties
  • Use in applications that do not involve high thermal stresses

Brown coloured to protect light-sensitive materials.

Specifications
  • Udv.Ø:
    47 mm
  • Kapacitet:
    100 ml
  • Højde:
    110 mm
  • Seq:
    04
Accessories

Product Information

  • Catalog #
    Beskrivelse
    Til
    Availability
    Price
  • 215-1876
    Screw cap, pipette and rubber suction bulb
    10 ml dropping bottles
    Log in for availability
    1 * 1 Items - 33,50 kr

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.