Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Dropping bottles, glass

Dropping bottles, glass

50 ml, 92 mm, 38 mm
Katalog # 215-3757
Dropping bottles, glass
Katalog # 215-3757
Leverandør:  
Dropping bottles, glass
Dropping bottles, glass

50 ml, 92 mm, 38 mm

Discontinued
Produktet er udgået. Men vi fandt lignende alternative produkter.
Alternativer
[EN]DROPPING BOTTLES AMBER 50 ML 1 * 144 Items Katalog #215-3821

Produktet 215-3821 ligner 215-3757, men producenten og specifikationerne kan variere.

[EN]PIPETTE KIT BLUE FOR 50ML BOTTLE 1 * 100 Items Katalog #215-3827

Produktet 215-3827 ligner 215-3757, men producenten og specifikationerne kan variere.

Product Details & Documents

Glass, amber, with screw cap, pipette and rubber suction bulb.

  • Good chemical and physical properties
  • Use in applications that do not involve high thermal stresses

Brown coloured to protect light-sensitive materials.

Specifikationer
  • Udv.Ø:
    38 mm
  • Kapacitet:
    50 ml
  • Højde:
    92 mm
  • Seq:
    03
Accessories

Produktinformation

  • Katalog #
    Til
    Tilgængelighed
    Price
  • Screw cap, pipette and rubber suction bulb
    Screw cap, pipette and rubber suction bulb
    215-1876
    10 ml dropping bottles
    Log ind for tilgængelighed
    1 * 1 Items - 33,50 kr