Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Dropping bottles, glass

Dropping bottles, glass

25 ml, 80 mm, 33 mm
Katalog # 215-3756
Dropping bottles, glass
Katalog # 215-3756
Leverandør:  
Dropping bottles, glass
Dropping bottles, glass

25 ml, 80 mm, 33 mm

Discontinued
Produktet er udgået. Men vi fandt lignende alternative produkter.
Alternativer
[EN]PIPETTE KIT BLUE FOR 30ML BOTTLE 1 * 100 Items Katalog #215-3826

Product 215-3826 is similar to 215-3756 but manufacturer and specifications may vary.

DROPPING BOTTLES AMBER 30 ML 1 * 110 Items Katalog #215-3820

Product 215-3820 is similar to 215-3756 but manufacturer and specifications may vary.

Product Details & Documents

Glass, amber, with screw cap, pipette and rubber suction bulb.

  • Good chemical and physical properties
  • Use in applications that do not involve high thermal stresses

Brown coloured to protect light-sensitive materials.

Specifikationer
  • Udv.Ø:
    33 mm
  • Kapacitet:
    25 ml
  • Højde:
    80 mm
  • Seq:
    02
Accessories

Produktinformation

  • Katalog #
    Til
    Tilgængelighed
    Price
  • Screw cap, pipette and rubber suction bulb
    Screw cap, pipette and rubber suction bulb
    215-1876
    10 ml dropping bottles
    Log ind for tilgængelighed
    1 * 1 Items - 33,50 kr