Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Infusion bottles

500 ml, GL 36, 76×188 mm
Katalog # 215-3763
Leverandør:   NORDIC PACK
Infusion bottles
Katalog # 215-3763
Leverandør:  NORDIC PACK
Infusion bottles
Infusion bottles

500 ml, GL 36, 76×188 mm

Price
5.800,00 kr
Log ind for tilgængelighed
1 * 35 Items
Product Details & Documents

Glass, clear.

  • Good chemical resistance
  • Good temperature resistance
  • Screw caps must be ordered seperately

Infusion bottles with thread.

Specifikationer
  • Diameter×H:
    76×188 mm
  • Gevind:
    GL 36
  • Kapacitet:
    500 ml
  • Seq:
    03