Orders LinkContactUsLinkComponent
Infusion bottles
Infusion bottles
-, GL 36, - mm
Katalog # 215-3764
Leverandør:   NORDIC PACK
Infusion bottles
Katalog # 215-3764
Leverandør:  NORDIC PACK
Infusion bottles
Infusion bottles
-, GL 36, - mm
Price
15,90 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Glass, clear.
  • Good chemical resistance
  • Good temperature resistance
  • Screw caps must be ordered seperately
Infusion bottles with thread.
Specifikationer
  • Diameter×H:
    - mm
  • Gevind:
    GL 36
  • Kapacitet:
    -
  • Seq:
    05