Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Infusion bottles

Infusion bottles

300 ml, GL 36, 65×160 mm
Katalog # 215-3767
Leverandør:   NORDIC PACK
Infusion bottles
Katalog # 215-3767
Leverandør:  NORDIC PACK
Infusion bottles
Infusion bottles

300 ml, GL 36, 65×160 mm

Price
97,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents

Glass, clear.

  • Good chemical resistance
  • Good temperature resistance
  • Screw caps must be ordered seperately

Infusion bottles with thread.

Specifikationer
  • Diameter×H:
    65×160 mm
  • Gevind:
    GL 36
  • Kapacitet:
    300 ml
  • Seq:
    02