Orders LinkContactUsLinkComponent
Infusion bottles
100 ml, GL 36, 47×118 mm
  215-3769
Infusion bottles
  215-3769
Infusion bottles
Infusion bottles
100 ml, GL 36, 47×118 mm
99,00 kr
1 * 1 Items
Glass, clear.
  • Good chemical resistance
  • Good temperature resistance
  • Screw caps must be ordered seperately
Infusion bottles with thread.
  • Diameter×H:
    47×118 mm
  • Gevind:
    GL 36
  • Kapacitet:
    100 ml
  • Seq:
    01