Orders LinkContactUsLinkComponent
Powder funnel, short stem
Powder funnel, short stem
100 mm, 26 mm, 34 mm
Katalog # 221-1160
Powder funnel, short stem
Katalog # 221-1160
Powder funnel, short stem
Powder funnel, short stem
100 mm, 26 mm, 34 mm
Price
137,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Soda-Kalkglas.
Specifikationer
  • Tragt udv. Ø:
    100 mm
  • Seq:
    07
  • Stilk udv. Ø:
    26 mm
  • Stilklængde:
    34 mm