Polyamidfilter, SUSP 70
55 mm
Katalog # 512-6149
Leverandør: Ahlstrom
Gå til:
Product Details & Documents
SUSP 70 is a woven polyamide fabric with a 70 μm mesh size used to capture coarse particles such as may be found in waste water.
Specifikationer
- Størrelse:55 mm