Orders LinkContactUsLinkComponent
Polyamidfilter, SUSP 70
Polyamidfilter, SUSP 70
50 mm
Katalog # 512-6150
Leverandør:   Ahlstrom
Polyamidfilter, SUSP 70
Katalog # 512-6150
Leverandør:  Ahlstrom
Polyamidfilter, SUSP 70
Polyamidfilter, SUSP 70
50 mm
Price
1.160,00 kr
Log ind for tilgængelighed
1 * 100 Items
Product Details & Documents
SUSP 70 is a woven polyamide fabric with a 70 μm mesh size used to capture coarse particles such as may be found in waste water.
Specifikationer
  • Størrelse:
    50 mm