Polyamidfilter, SUSP 70
47 mm
Katalog # 512-6151
Leverandør: Ahlstrom
Gå til:
Product Details & Documents
SUSP 70 is a woven polyamide fabric with a 70 μm mesh size used to capture coarse particles such as may be found in waste water.
Specifikationer
- Størrelse:47 mm