Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

High purity qualitative filter paper

5, 320 mm
Catalog # 512-6081
Supplier:   Ahlstrom
High purity qualitative filter paper
Catalog # 512-6081
Supplier:  Ahlstrom
High purity qualitative filter paper
High purity qualitative filter paper

5, 320 mm

Price
564,00 kr
Log in for availability
1 * 50 Items
Product Details & Documents

Ahlstrom-Munksjö’s qualitative grades of filter paper are recommended for use in analytical methods which determine or identify particulate constituents of a mixture irrespective of the amount present. Qualitative filter papers are often used in routine separation work that still requires high purity and consistent performance. These filter papers covers a wide range of laboratory applications, as liquids clarification, qualitative analytical separations for precipitates and buffers filtration, and are also used for soil analysis and in food and beverage testing. Wet-strengthened grades contain a reinforcement agent that increases their resistance to rupture when wet, such as in vacuum filtration, and their resistance when filtering acidic solutions.

Grade 5 and V5: Very fast filtering, open fiber matrix. For coarse and voluminous precipitates such as iron- aluminum- and chromium hydroxide; Si- determination in steel and pig iron analysis.
Grade 3: Fast filtering, open fiber matrix. For coarse and voluminous precipitates such as iron- aluminum- and chromium hydroxide; Si- determination in steel and pig iron analysis.

Grade V150: Medium fast filtering, open to medium fiber matrix. For qualitative tasks, coarser precipitates such as lead-, iron- and silver sulfide; alkali carbonates; calcium oxalate.
Grade 1F and V1F: Medium to slowly filtering, medium tight fiber matrix. For fast filtration of fine precipitates such as calcium oxalate, magnesium ammonium phosphate, coarser forms of barium sulphate.

Grade Munktell no. 1 and 292: Medium to slowly filtering, medium tight fiber matrix. For fast filtration of fine precipitates such as calcium oxalate, magnesium ammonium phosphate, coarser forms of barium sulfate.
Grade Munktell no. 2 and 292a: Slowly filtering, tight fiber matrix. For filtration of fine precipitates such as lead dioxide, calcium fluoride, nickel sulfide, zinc sulfide.

Grade 106: Slowly filtering for fine precipitates such as lead dioxide, calcium fluoride, nickel sulphide and zinc sulphide.
Grade 120H and V120H: Very slowly filtering, tight fiber matrix. For fine precipitates such as barium sulphate, metastannic acid and cuprous oxide.

Specifications
  • Kvalitet:
    5
  • Seq:
    58
  • Størrelse:
    320 mm
  • Filtreringshastighed:
    60 s (Herzberg)
  • Cat. No.:
    512-6081
  • Partikelretention:
    20 µm
  • Vægt:
    130 g/m²
  • Askeindhold:
    0,06%
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.