Cheers, tray
180×240×17 mm
Katalog # 216-0423
Leverandør: VITLAB
Gå til:
Product Details & Documents
Made of melamine, white.
- Rounded corners
- Smooth surfaces, easy to clean
- Stable and self-supporting
Practical tray for instruments, tools, and sensitive utensils.
Specifikationer
- B×D×H:180×240×17 mm