Screw Caps
GL 40
Katalog # 215-2802
Leverandør: GLASWARENFABRIK KARL HECHT
Gå til:
Product Details & Documents
PP, sort.
Very good chemical resistance
- Heat-resistant up to 110 °C
- Only suitable for bottles 215-2814 to 215-2819
Specifikationer
- Gevind:GL 40
- Seq:5