PTFE balls, BOLA
Balls, PTFE, 3 mm
Katalog # BOHLH964-03
Leverandør: Bohlender
Gå til:
Product Details & Documents
Made from solid PTFE, with smooth surface.
For extension of surfaces of e.g. distillation apparatus; as splash guard.
Specifikationer
- Ø:3 mm
- Beskrivelse:Balls
- Materiale:PTFE
- Seq:002