Orders LinkContactUsLinkComponent
PTFE balls, BOLA
Balls, PTFE, 3 mm
Katalog # BOHLH964-03
Leverandør:   Bohlender
PTFE balls, BOLA
Katalog # BOHLH964-03
Leverandør:  Bohlender
PTFE balls, BOLA
PTFE balls, BOLA
Balls, PTFE, 3 mm
Price
1.440,00 kr
Log ind for tilgængelighed
1 * 100 Items
Product Details & Documents
Made from solid PTFE, with smooth surface.
For extension of surfaces of e.g. distillation apparatus; as splash guard.
Specifikationer
  • Ø:
    3 mm
  • Beskrivelse:
    Balls
  • Materiale:
    PTFE
  • Seq:
    002