Natriumdodecylsulfat (SDS)
Glass bottle for solids, 100 g
Katalog # RESERES9010L-A102X
Leverandør: RESEARCH ORGANICS
Natriumdodecylsulfat (SDS)
Katalog # RESERES9010L-A102X
Leverandør: RESEARCH ORGANICS
CAS nummer: 151-21-3
Gå til:
Product Details & Documents
Synonymer: Dodecyl sulphate sodium salt, Sodium dodecyl sulfate, Dodecyl sulphuric acid sodium salt, Lauryl sulphate sodium salt, Sodium lauryl sulphate
- CAS nummer:151-21-3
- Formel:CH₃(CH₂)₁₁OSO₃Na
- MDL nummer:MFCD00036175
- Molecular Weight:288.38 g/mol
- Merck Index:14,08636
- Smeltende Pt:204…207 °C
- Tæthed:1,1 g/cm³ (20 °C)
- Flash Pt:>100 °C
- Opbevaringstemperatur:Stue temperatur
- EINECS:205-788-1
- ADR:4.1,II
- :M-Clarity™ quality level = MQ300
- FN:1325
Specifikationer
Specifikationer
- Pakning:Glass bottle for solids
- Pakn:100 g
Specifikationer
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



