Natriumdodecylsulfat (SDS)
Glass bottle for solids, 1 kg
RESERES9010L-A103X
: Dodecyl sulphate sodium salt, Sodium dodecyl sulfate, Dodecyl sulphuric acid sodium salt, Lauryl sulphate sodium salt, Sodium lauryl sulphate
- :151-21-3
- :CH₃(CH₂)₁₁OSO₃Na
- :MFCD00036175
- :288.38 g/mol
- :14,08636
- :204…207 °C
- :1,1 g/cm³ (20 °C)
- :>100 °C
- :Stue temperatur
- :205-788-1
- :4.1,II
- :M-Clarity™ quality level = MQ300
- :1325
- Pakning:Glass bottle for solids
- Pakn:1 kg