Orders LinkContactUsLinkComponent
Natriumdodecylsulfat (SDS)
Glass bottle for solids, 1 kg
  RESERES9010L-A103X
Natriumdodecylsulfat (SDS)
  RESERES9010L-A103X
 :  151-21-3
Natriumdodecylsulfat (SDS)
Natriumdodecylsulfat (SDS)
Glass bottle for solids, 1 kg
 :  151-21-3
1 * 1 kg
 :  Dodecyl sulphate sodium salt, Sodium dodecyl sulfate, Dodecyl sulphuric acid sodium salt, Lauryl sulphate sodium salt, Sodium lauryl sulphate
  •  :151-21-3
  •  :CH₃(CH₂)₁₁OSO₃Na
  •  :MFCD00036175
  •  :288.38 g/mol
  •  :14,08636
  •  :204…207 °C
  •  :1,1 g/cm³ (20 °C)
  •  :>100 °C
  •  :Stue temperatur
  •  :205-788-1
  •  :4.1,II
  •  :M-Clarity™ quality level = MQ300
  •  :1325
  • Pakning:
    Glass bottle for solids
  • Pakn:
    1 kg