Orders LinkContactUsLinkComponent
Tricine
Glass bottle for solids, 100 g
  RESERES3077T-A102X
Tricine
  RESERES3077T-A102X
 :  5704-04-1
Tricine
Glass bottle for solids, 100 g
 :  5704-04-1
1 * 100 g
 :  2-((1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl)amino)acetic acid, Tricine buffer, N-(Tris(hydroxymethyl)methyl)glycine
  •  :5704-04-1
  •  :(HOCH₂)₃CNHCH₂CO₂H
  •  :MFCD00004277
  •  :179.17 g/mol
  •  :13,09724
  •  :187 °C
  •  :1,523 g/cm³ (19.85 °C)
  •  :Stue temperatur
  •  :227-193-6
  • Pakning:
    Glass bottle for solids
  • Pakn:
    100 g