Tricine
Glass bottle for solids, 100 g
RESERES3077T-A102X
: 2-((1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl)amino)acetic acid, Tricine buffer, N-(Tris(hydroxymethyl)methyl)glycine
- :5704-04-1
- :(HOCH₂)₃CNHCH₂CO₂H
- :MFCD00004277
- :179.17 g/mol
- :13,09724
- :187 °C
- :1,523 g/cm³ (19.85 °C)
- :Stue temperatur
- :227-193-6
- Pakning:Glass bottle for solids
- Pakn:100 g