Tricine
Glass bottle for solids, 1 kg
Katalog # RESERES3077T-A103X
Leverandør: RESEARCH ORGANICS
Gå til:
Product Details & Documents
Synonymer: 2-((1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl)amino)acetic acid, Tricine buffer, N-(Tris(hydroxymethyl)methyl)glycine
- CAS nummer:5704-04-1
- Formel:(HOCH₂)₃CNHCH₂CO₂H
- MDL nummer:MFCD00004277
- Molecular Weight:179.17 g/mol
- Merck Index:13,09724
- Smeltende Pt:187 °C
- Tæthed:1,523 g/cm³ (19.85 °C)
- Opbevaringstemperatur:Stue temperatur
- EINECS:227-193-6
Specifikationer
Specifikationer
- Pakning:Glass bottle for solids
- Pakn:1 kg
Specifikationer
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



