MOPS
Glass bottle for solids, 250 g
Katalog # RESERES0195M-A103X
Leverandør: RESEARCH ORGANICS
Gå til:
Product Details & Documents
Synonymer: 3-Morpholinopropane-1-sulphonic acid, 3-(N-Morpholino)propanesulphonic acid
- CAS nummer:1132-61-2
- Formel:C₇H₁₅NO₄S
- MDL nummer:MFCD00006183
- Molecular Weight:209.27 g/mol
- Merck Index:13,06288
- Smeltende Pt:> 250 °C
- Flash Pt:110 °C (closed cup)
- Opbevaringstemperatur:Stue temperatur
- EINECS:214-478-5
Specifikationer
Specifikationer
- Pakning:Glass bottle for solids
- Pakn:250 g
Specifikationer
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



