MOPS
Glass bottle for solids, 25 g
RESERES0195M-A101X
: 3-Morpholinopropane-1-sulphonic acid, 3-(N-Morpholino)propanesulphonic acid
- :1132-61-2
- :C₇H₁₅NO₄S
- :MFCD00006183
- :209.27 g/mol
- :13,06288
- :> 250 °C
- :110 °C (closed cup)
- :Stue temperatur
- :214-478-5
- Pakning:Glass bottle for solids
- Pakn:25 g
- Cat. No.:RESERES0195M-A101X
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



