MOPS
Glass bottle for solids, 100 g
Katalog # RESERES0195M-A102X
Leverandør: RESEARCH ORGANICS
Gå til:
Product Details & Documents
Synonymer: 3-Morpholinopropane-1-sulphonic acid, 3-(N-Morpholino)propanesulphonic acid
- CAS nummer:1132-61-2
- Formel:C₇H₁₅NO₄S
- MDL nummer:MFCD00006183
- Molecular Weight:209.27 g/mol
- Merck Index:13,06288
- Smeltende Pt:> 250 °C
- Flash Pt:110 °C (closed cup)
- Opbevaringstemperatur:Stue temperatur
- EINECS:214-478-5
Specifikationer
Specifikationer
- Pakning:Glass bottle for solids
- Pakn:100 g



