MOPS
Glass bottle for solids, 1 kg
Catalog # RESERES0195M-A104X
Supplier: RESEARCH ORGANICS
Jump to:
Product Details & Documents
Synonyms: 3-Morpholinopropane-1-sulphonic acid, 3-(N-Morpholino)propanesulphonic acid
- CAS Number:1132-61-2
- Formula:C₇H₁₅NO₄S
- MDL Number:MFCD00006183
- Molecular Weight:209.27 g/mol
- Merck Index:13,06288
- Melting Pt:> 250 °C
- Flash Pt:110 °C (closed cup)
- Storage Temperature:Stue temperatur
- EINECS:214-478-5
Specifications
Specifications
- Pakning:Glass bottle for solids
- Pakn:1 kg



