Stivelse (from potato)
Glass bottle for solids, 1 kg
Katalog # ROTH9441.1
Marketsource
Leverandør: Roth Carl
Gå til:
Product Details & Documents
- CAS nummer:9005-25-8
- Formel:(C₆H₁₀O₅)n
- MDL nummer:MFCD00082026
- Molecular Weight:162.14 g/mol
- Opbevaringstemperatur:Stue temperatur
- EINECS:232-679-6
Specifikationer
Specifikationer
- Pakning:Glass bottle for solids
- Pakn:1 kg
Specifikationer
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



