Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Zink, powder

Zink, powder

Glass bottle for solids, 500 g
Katalog # ROTH9524.2
Brandfarlig
Miljø
Marketsource
Leverandør:   Roth Carl
Zink, powder
Katalog # ROTH9524.2
Leverandør:  Roth Carl
Marketsource
CAS nummer:  7440-66-6
Zink, powder

Glass bottle for solids, 500 g

CAS nummer:  7440-66-6
Price
Spørg for pris
Log ind for tilgængelighed
1 * 500 g
Product Details & Documents
  • CAS nummer:7440-66-6
  • Formel:Zn
  • MDL nummer:MFCD00011291
  • Molecular Weight:65.39 g/mol
  • Kogende Pt:907 °C (1013 hPa)
  • Smeltende Pt:419,5 °C
  • Tæthed:7,13 g/cm³ (20 °C)
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:231-175-3
  • FN:1436
  • ADR:4.3,II
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle for solids
  • Pakn:
    500 g

Specifikationer

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.