Kolophonium
Glass bottle for solids, 1 kg
Katalog # ROTH9051.1
Marketsource
Leverandør: Roth Carl
Gå til:
Product Details & Documents
Synonymer: Terpentinoliefri harpiks
- CAS nummer:8050-09-7
- MDL nummer:MFCD00132205
- Smeltende Pt:75…81 °C
- Tæthed:1,1 g/cm³ (20 °C)
- Flash Pt:188 °C
- EINECS:232-475-7
Specifikationer
Specifikationer
- Pakning:Glass bottle for solids
- Pakn:1 kg