Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Antimon, powder

Antimon, powder

Glass bottle for solids, 100 g
Katalog # ROTH9258.1
Irriterende
Miljø
Marketsource
Leverandør:   Roth Carl
Antimon, powder
Katalog # ROTH9258.1
Leverandør:  Roth Carl
Marketsource
CAS nummer:  7440-36-0
Antimon, powder

Glass bottle for solids, 100 g

CAS nummer:  7440-36-0
Price
Spørg for pris
Log ind for tilgængelighed
1 * 100 g
Product Details & Documents
  • CAS nummer:7440-36-0
  • Formel:Sb
  • MDL nummer:MFCD00134030
  • Molecular Weight:121.76 g/mol
  • EINECS:231-146-5
  • FN:2871
  • ADR:6.1,III
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle for solids
  • Pakn:
    100 g

Specifikationer

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.