Orders LinkContactUsLinkComponent
Conical separating funnels
Conical separating funnels
50 ml, 14/23 NS
Katalog # 532-0103
Leverandør:   SciLabware
Conical separating funnels
Katalog # 532-0103
Leverandør:  SciLabware
Conical separating funnels
Conical separating funnels
50 ml, 14/23 NS
Price
929,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
PYREX®, borosilicate glass, with glass stopper.
With ground glass stopper and interchangeable PTFE key. Without graduation.
Specifikationer
  • Kappestr.:
    14/23 NS
  • Kapacitet:
    50 ml
  • Seq:
    01