Dækglas, Menzel-Gläser
Cover glasses, 24×60 mm, #2
Catalog # 630-2895
Supplier: Epredia
Jump to:
Product Details & Documents
These cover slips are suitable for in vitro diagnostic (IVD) applications.
- Uniform quality of thickness
- No bubbles or flaws
- Made of pre-washed pure white Hydrolytic Class 1 glass
Documents
Specifications
- Antal:#2
- Beskrivelse:Cover glasses
- L×B:24×60 mm
- Seq:33