Dækglas, Menzel-Gläser
24×24 mm, #1
: Epredia
Pre-washed pure white D 263 M borosilicate glass of hydrolytic class 1 cover slips.
- High spectral transmission, uniform quality of thickness
- No bubbles or flaws guarantees high optical homogeneity
- Highly transparent and achromatic: Practically free of any colour distortion
Suitable for in vitro diagnostic (IVD) applications in medicine, biology and research examinations.
: DIN ISO 719 ; DIN ISO 12 116 ; DIN ISO 695.
- Antal:#1
- L×B:24×24 mm
- Seq:34