Dækglas, Menzel-Gläser
Cover glasses, 22×32 mm, #1,5
630-2879
: Epredia
These cover slips are suitable for in vitro diagnostic (IVD) applications.
- Uniform quality of thickness
- No bubbles or flaws
- Made of pre-washed pure white Hydrolytic Class 1 glass
Documents
- Antal:#1,5
- Beskrivelse:Cover glasses
- L×B:22×32 mm
- Seq:08