Orders LinkContactUsLinkComponent
Dækglas, Menzel-Gläser
20×20 mm, #1,5
Katalog # 630-2886
Leverandør:   Epredia
Dækglas, Menzel-Gläser
Katalog # 630-2886
Leverandør:  Epredia
Dækglas, Menzel-Gläser
Dækglas, Menzel-Gläser
20×20 mm, #1,5
Price
298,00 kr
Log ind for tilgængelighed
1 * 1.000 Items
Product Details & Documents
Pre-washed pure white D 263 M borosilicate glass of hydrolytic class 1 cover slips.
  • High spectral transmission, uniform quality of thickness
  • No bubbles or flaws guarantees high optical homogeneity
  • Highly transparent and achromatic: Practically free of any colour distortion
Suitable for in vitro diagnostic (IVD) applications in medicine, biology and research examinations.
 : DIN ISO 719 ; DIN ISO 12 116 ; DIN ISO 695.
Specifikationer
  • Antal:
    #1,5
  • L×B:
    20×20 mm
  • Seq:
    20