Dækglas, Menzel-Gläser
18×18 mm, #1,5
Leverandør: Epredia
Gå til:
Product Details & Documents
Pre-washed pure white D 263 M borosilicate glass of hydrolytic class 1 cover slips.
- High spectral transmission, uniform quality of thickness
- No bubbles or flaws guarantees high optical homogeneity
- Highly transparent and achromatic: Practically free of any colour distortion
Suitable for in vitro diagnostic (IVD) applications in medicine, biology and research examinations.
Certifications: DIN ISO 719 ; DIN ISO 12 116 ; DIN ISO 695.
Documents
Certifikater
Søg efter partinummerSpecifikationer
- Antal:#1,5
- L×B:18×18 mm
- Seq:15