Orders LinkContactUsLinkComponent
Dental forceps, serrated
Dental forceps, serrated
College, bent, serrated, 160 mm
Katalog # HAMMHTC034-05
Leverandør:   HAMMACHER KARL
Dental forceps, serrated
Katalog # HAMMHTC034-05
Leverandør:  HAMMACHER KARL
Dental forceps, serrated
Dental forceps, serrated
College, bent, serrated, 160 mm
Price
745,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Dental forceps made from titanium.
Specifikationer
  • Model:
    College, bent, serrated
  • Længde:
    160 mm
  • Seq:
    02