Pipettespidser, universal, Axygen®
Extended, 0,5 - 10 µl, -, Transparent, Stack racks, 96 pr. stk.
613-0644
: Corning
Disse universelle pipettespidser er Kompatibel med de fleste mikropipetter.
- PP, autoclavable
- Smooth finish for minimum retention of collected sample
- Certified free of RNase, DNase and endotoxins
- Maxymum Recovery® tips feature ultra smooth surfaces: For reduced sample loss and maximal accuracy and precision
: Fås løse eller i racks, sterile eller usterile versioner.
- Pakket:Stack racks, 96 pr. stk.
- Steril:-
- Volumen:0,5 - 10 µl
- Seq:08
- Spidstype:Extended
- Spits farge:Transparent