Pipettespidser, universal, Axygen®
Bevelled, 1 - 200 µl, -, Yellow, Stack Rack inkl. 5 racks, 96 pr. stk.
Katalog # 613-1291
Leverandør: Corning
Gå til:
Product Details & Documents
Disse universelle pipettespidser er Kompatibel med de fleste mikropipetter.
- PP, autoclavable
- Smooth finish for minimum retention of collected sample
- Certified free of RNase, DNase and endotoxins
- Maxymum Recovery® tips feature ultra smooth surfaces: For reduced sample loss and maximal accuracy and precision
: Fås løse eller i racks, sterile eller usterile versioner.
Specifikationer
- Pakket:Stack Rack inkl. 5 racks, 96 pr. stk.
- Steril:-
- Volumen:1 - 200 µl
- Seq:21
- Spidstype:Bevelled
- Spits farge:Yellow