Orders LinkContactUsLinkComponent
Pipettespidser, universal, Axygen®
Bevelled, 1 - 200 µl, -, Yellow, Stack Rack inkl. 5 racks, 96 pr. stk.
Katalog # 613-1291
Leverandør:   Corning
Pipettespidser, universal, Axygen®
Katalog # 613-1291
Leverandør:  Corning
Pipettespidser, universal, Axygen®
Pipettespidser, universal, Axygen®
Bevelled, 1 - 200 µl, -, Yellow, Stack Rack inkl. 5 racks, 96 pr. stk.
Price
210,00 kr
Log ind for tilgængelighed
1 * 480 Items
Product Details & Documents
Disse universelle pipettespidser er Kompatibel med de fleste mikropipetter.
  • PP, autoclavable
  • Smooth finish for minimum retention of collected sample
  • Certified free of RNase, DNase and endotoxins
  • Maxymum Recovery® tips feature ultra smooth surfaces: For reduced sample loss and maximal accuracy and precision
 : Fås løse eller i racks, sterile eller usterile versioner.
Specifikationer
  • Pakket:
    Stack Rack inkl. 5 racks, 96 pr. stk.
  • Steril:
    -
  • Volumen:
    1 - 200 µl
  • Seq:
    21
  • Spidstype:
    Bevelled
  • Spits farge:
    Yellow